Platinum rhodium alloy (90:10 w%) ≥99.9% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
MDL Number:MFCD00198186
Storage Temperature:Ambient
Related Information
Frequently Bought Together