Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Agar

Agar

Glass bottle for solids, 500 g
Catalog # A3477.0500
Supplier:   PanReac AppliChem
Agar
Catalog # A3477.0500
Supplier:  PanReac AppliChem
CAS Number:  9002-18-0
Agar
Agar

Glass bottle for solids, 500 g

CAS Number:  9002-18-0
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.