L(-)-Glucose, anhydrous 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:921-60-8
:
Formula:C₆H₁₂O₆
MDL Number:MFCD00148913
:180.16 g/mol
Boiling Pt:376 °C (1013 hPa)
Melting Pt:148…157 °C
Storage Temperature:Ambient
EINECS:213-068-3
Related Information
Frequently Bought Together