
L(+)-Lactic acid 20% (w/w) in water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79-33-4
:
Formula:C₃H₆O₃
MDL Number:MFCD00064266
:90.08 g/mol
Density:1,05 g/cm³
EINECS:201-196-2
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Solution
- Titration with NaOH:≥18 %
- Sulfate (SO4):≤30 ppm
- Iron (Fe):≤10 ppm
Specification Test Results
Frequently Bought Together