D-(+)-Trehalose dihydrate
:6138-23-4
:
:C₁₂H₂₂O₁₁·2H₂O
:MFCD00071594
:378.33 g/mol
:97…99 °C
:Ambient
:202-739-6
Frequently Bought Together
VWR®, Screw Caps for Short Thread Vials ND9
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Standard Line Transfer Pipettes
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Deep well and assay plates, Axygen®
: Corning
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
Charcoal activated, pellets, Norit® ROW 0.8 mm, steam activated
: Thermo Fisher Scientific