Tungsten hexacarbonyl for synthesis, Sigma-Aldrich®
Catalog # 8.00888.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:150 °C
- Molar Mass:351.91 g/mol
- Boiling Point:175 °C (1013 hPa)
- Vapor Pressure:1.4 hPa (67 °C)
- Density:2.65 g/cm3 (20 °C)
- Synonyms:Tungsten carbonyl
- Hill Formula:C6O6W
- Chemical Formula:W(CO)6
Specification Test Results
About this item
:
:351.9 g/mol
Boiling Pt:451,3 °C (1013 hPa)
Melting Pt:169…171 °C
Density:2,65 g/cm³ (20 °C)
EINECS:237-880-2
UN:3466
ADR:6.1,III
:M-Clarity™ quality level = MQ200
CAS Number:14040-11-0
Formula:C₆O₆W
MDL Number:MFCD00011462