Order Entry
Northern Ireland
ContactUsLinkComponent
Dimethyl sulphate-[D₆] 99+ atom % D for NMR spectroscopy
Toxic
Health
Corrosive
Dimethyl sulphate-[D₆] 99+ atom % D for NMR spectroscopy
Catalog # ACRO172910010
CAS Number:  
undefined
Toxic
Health
Corrosive
Dimethyl sulphate-[D₆] 99+ atom % D for NMR spectroscopy
Catalog # ACRO172910010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.05 %
  • D-Enrichment:
    ≥99.0 %

Specification Test Results

About this item

 :
 :132.09 g/mol
Boiling Pt:60…61 °C (7 mmHg)
Melting Pt:–32 °C
Density:1,39 g/cm³
Flash Pt:83 °C
Storage Temperature:Ambient
EINECS:239-257-0
UN:1595
ADR:6.1,I
 :GHS06
 :GHS08
 :GHS05
CAS Number:15199-43-6
Formula:D₃CS(O)₂CD₃