Molecular sieve A5 (0.5 nm, 5 Å), pellets, Ø 1.6 mm, Fluka™
Catalog # 334316-1KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
Specifications
Specification Test Results
- Particle Size:1.6 mm
- Form:pellets
- Granulation:complying
- Pore Size (Ång Ström):5
Specification Test Results
About this item
:
CAS Number:69912-79-4
:1318-02-1