Order Entry
Northern Ireland
ContactUsLinkComponent
Molecular sieve 13X (1.0 nm, 10 Å), Supelco®
Molecular sieve 13X (1.0 nm, 10 Å), Supelco®
Catalog # 1.05703.1000
Supplier:  Merck
CAS Number:  
Molecular sieve 13X (1.0 nm, 10 Å), Supelco®
Catalog # 1.05703.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Bulk Density:
    700 - 750 kg/m3
  • Synonyms:
    Sodium aluminium silicate

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:215-283-8
CAS Number:63231-69-6
 :1318-02-1
MDL Number:MFCD00164506