
Isoeugenol cis- and trans-mixture 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:97-54-1
:
Formula:C₁₀H₁₂O₂
MDL Number:MFCD00009285
Merck Index:13,05189
Boiling Pt:266 °C (1013 hPa)
Melting Pt:19…23 °C
Density:1,09 g/cm³
Flash Pt:150 °C
:164.2 g/mol
EINECS:202-590-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.5750 to 1.5790 (20°C, 589 nm)
- GC:≥98.0 %
Specification Test Results
Frequently Bought Together