1,5-Cyclooctadiene 99% stabilised
:111-78-4
:
:C₈H₁₂
:MFCD00001752
:108.18 g/mol
:151 °C (101,3kPa)
:–69,5 °C
:0,88 g/cm³
:38 °C
:203-907-1
:2520
:3,III
:GHS07
:GHS02
:GHS08
:GHS09
:Ambient
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4920 to 1.4950 (20°C, 589 nm)
- GC:≥98.5 %
- Stabilizer:100 to 200 ppm Octadecyl 3-(3',5'-di-tert-butyl
Frequently Bought Together