ContactUsLinkComponent
Eyewash station, Combibox DUO
Eyewash station, Combibox DUO
Catalog # PLUM4862
Supplier:  Plum Safety
undefined
Eyewash station, Combibox DUO
Catalog # PLUM4862
Supplier:  Plum Safety
Specifications
  • Description:
    Eyewash station, Combibox DUO, incl. 500 ml pH Neutral DUO and 500 ml Plum eye wash DUO
  • Pk:
    1
Specifications
About this item
Wall-mounted eye wash box made from maintenance-free material for quick first aid of eye injuries. The box contains one 500 ml bottle of pH Neutral DUO and one 500 ml bottle of Plum eye wash DUO.
  • Dust-tight wall station in high-impact PS
  • Quick and efficient eye wash
  • Quick and easy to mount on the wall close to the workplace
  • With clear pictograms and mirror
  • Particularly suited for dusty and dirty production areas
W×D×H: 230×110×280 mm