Order Entry
Northern Ireland
ContactUsLinkComponent
 
Platinum, black 98%
 
undefined
Platinum, black 98%

 

  • Appearance:
    Black Powder
  • Assay:
    ≥97.5 % (Pt)
  • Specific surface area:
    ≥25 m²/g

 

 

Frequently Bought Together