Iridium ≥99.8% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-88-5
:
Formula:Ir
MDL Number:MFCD00011062
:192.22 g/mol
Boiling Pt:5300 °C (1013 hPa)
Melting Pt:2450 °C
Storage Temperature:Ambient
EINECS:231-095-9
Frequently Bought Together