Iron ≥99.995% (metals basis), foil, Puratronic®, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-89-6
:
Formula:Fe
MDL Number:MFCD00010999
Boiling Pt:3000
Melting Pt:1535
Density:7.86 g/cm³ (20 °C)
Molecular Weight:55.85 g/mol
Merck Index:13,05109
Storage Temperature:Ambient
EINECS:231-096-4
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together