Iron ≥99.95% (metals basis), slug, Length: 9.5 mm (0.37 in), Ø 9.5 mm (0.37 in)
Catalog # 00769.18
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:55.85 g/mol
Merck Index:13,05109
Boiling Pt:3000 °C (1013 hPa)
Melting Pt:1535 °C
Density:7,86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-096-4
CAS Number:7439-89-6
Formula:Fe
MDL Number:MFCD00010999