ContactUsLinkComponent
Cobalt ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Cobalt ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
  40834.KS
 :  
Cobalt ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
  40834.KS
 :  
  • Pk:
    1
  • Seq:
    0001
 :7440-48-4
 :
 :Co
 :MFCD00010935
 :58.93 g/mol
 :12,02488
 :2870 °C (1013 hPa)
 :1495 °C
 :8,9 g/cm³ (20 °C)
 :231-158-0
 :GHS08
 :Ambient