Zirconium ≥99.9% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-67-7
:
Formula:Zr
MDL Number:MFCD00011303
:91.22 g/mol
Storage Temperature:Ambient
EINECS:231-176-9
Related Information
Frequently Bought Together