Order Entry
Northern Ireland
ContactUsLinkComponent
Zirconium ≥99.2% (metals basis), sponge ≤ 25.4 mm (1.0 in)
Flammable
Zirconium ≥99.2% (metals basis), sponge ≤ 25.4 mm (1.0 in)
Catalog # 14191.36
CAS Number:  
Flammable
Zirconium ≥99.2% (metals basis), sponge ≤ 25.4 mm (1.0 in)
Catalog # 14191.36
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
ADR:4.2,III
 :91.22 g/mol
EINECS:231-176-9
UN:2008
 :GHS02
Storage Temperature:Ambient
CAS Number:7440-67-7
Formula:Zr
MDL Number:MFCD00011303