Yttrium ≥99.9% (REO, rare earth oxide basis), ingot
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-65-5
:
Formula:Y
MDL Number:MFCD00011468
:88.91 g/mol
Merck Index:12,10240
Boiling Pt:2927 °C (1013 hPa)
Melting Pt:1525 °C
Density:4,472 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-174-8
Related Information
Frequently Bought Together