Silver ≥99.9% (metals basis), spherical powder -500 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-22-4
:
Formula:Ag
MDL Number:MFCD00003397
Boiling Pt:2212
Melting Pt:962
Density:10.5 g/cm³ (20 °C)
Molecular Weight:107.87 g/mol
Storage Temperature:Ambient
EINECS:231-131-3
ADR:9 III
Merck Index:12,08647
UN:3077
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together