
Samarium for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7440-19-9
:
Formula:Sm
MDL Number:MFCD00011233
:150.36 g/mol
Merck Index:12,08498
Boiling Pt:1900 °C (1013 hPa)
Melting Pt:1076,85 °C
Density:7,42 g/cm³ (20 °C)
UN:3208
ADR:4.3,I
:M-Clarity™ quality level = MQ200
EINECS:231-128-7
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Melting Point:1072 °C
- Molar Mass:150.36 g/mol
Specification Test Results
Frequently Bought Together