Order Entry
Northern Ireland
ContactUsLinkComponent
Samarium ≥99.5% (REO, rare earth oxide basis), ingot/button, Ø ∼50.8 mm (2.0 in)
Samarium ≥99.5% (REO, rare earth oxide basis), ingot/button, Ø ∼50.8 mm (2.0 in)
Catalog # 45107.KF
CAS Number:  
Samarium ≥99.5% (REO, rare earth oxide basis), ingot/button, Ø ∼50.8 mm (2.0 in)
Catalog # 45107.KF
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7440-19-9
 :
Formula:Sm
MDL Number:MFCD00011233
 :150.36 g/mol
Merck Index:12,08498
Boiling Pt:1900 °C (1013 hPa)
Melting Pt:1076,85 °C
Density:7,42 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-128-7