
Samarium ≥99.9% (REO, rare earth oxide basis), pieces, sublimed dendritic
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-19-9
:
Formula:Sm
MDL Number:MFCD00011233
:150.36 g/mol
Merck Index:12,08498
Boiling Pt:1900 °C (1013 hPa)
Melting Pt:1076,85 °C
Density:7,42 g/cm³ (20 °C)
UN:3178
ADR:4.1,III
:Packaged under argon.
EINECS:231-128-7
:GHS02
Storage Temperature:Ambient
Related Information
Frequently Bought Together