Order Entry
Northern Ireland
ContactUsLinkComponent
Bismuth 99.5%, powder -100 mesh
Flammable
Bismuth 99.5%, powder -100 mesh
Certificates
undefined
Flammable
Bismuth 99.5%, powder -100 mesh

Specification Test Results

  • Appearance:
    Golden to brown to grey Powder
  • Particle size:
    ≤100 mesh
  • Total trace metal impurities:
    ≤0.5 %

Specification Test Results

Related Information

Frequently Bought Together