Tubes, quartz
Supplier: TECHNICAL QUARTZ SOLUTIONS
Certificates
About this item
Fused silica tubes are made from natural quartz to ensure high quality and versatility
- With an operating range of up to 1200 °C
- Very good chemical resistance
Frequently Bought Together