
Glassy carbon, spherical powder 630-1000 µm Type 1
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-44-0
:
Formula:C
MDL Number:MFCD00133992
:12.01 g/mol
Merck Index:13,01818
Melting Pt:3652 °C
Density:1,95 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-153-3
UN:1325
ADR:4.1,III
:GHS02
Frequently Bought Together