
Glassy carbon, powder 0,4-12 µm Type2
Catalog # 38007.18
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:12.01 g/mol
Merck Index:13,01818
Melting Pt:3652 °C
Density:1,95 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-153-3
UN:1325
ADR:4.1,III
:GHS02
CAS Number:7440-44-0
Formula:C
MDL Number:MFCD00133992