Carbon ≥99%, felt, Thickness: 6.35 mm (0.250 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-44-0
:
Formula:C
MDL Number:MFCD00133992
:12.01 g/mol
Merck Index:13,01818
Boiling Pt:∼4000 °C (1013 hPa)
Melting Pt:3727 °C
Density:3,65 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-955-3
Related Information
Frequently Bought Together