Carbon ≥99.999% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 40832.KS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:12.01 g/mol
Merck Index:13,01818
Boiling Pt:∼4000 °C (1013 hPa)
Melting Pt:3727 °C
Density:3,65 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-955-3
CAS Number:7440-44-0
Formula:C
MDL Number:MFCD00133992