Aluminium ≥99.999% (metals basis), slug, Puratronic®, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7429-90-5
:
Formula:Al
MDL Number:MFCD00134029
:26.98 g/mol
Merck Index:13,00321
Boiling Pt:2327 °C (1013 hPa)
Melting Pt:660 °C
Density:2,7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-072-3
Frequently Bought Together