Order Entry
Northern Ireland
ContactUsLinkComponent
 
Ointment jar
  215-3318
 :  Azpack
undefined
Ointment jar
  215-3318
 :  Azpack

 

  • Cap Size:
    R3/58
  • Ext.Ø:
    62,30 mm
  • Capacity:
    120 ml
  • Height:
    63,50 mm
  • Pk:
    20

 

 

These amber ointment jars made from type III soda lime glass fitted with R3/58 black urea cap and woodpulp p.v.d.c faced liner.

  • Good chemical resistance
  • Good temperature resistance

Amber coloured to protect light sensitive substances.