Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
HC Toxin

HC Toxin

1 mg
Catalog # ALEXBML-GR320-0001
HC Toxin
Catalog # ALEXBML-GR320-0001
Supplier:  
CAS Number:  
HC Toxin

1 mg

CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

HDAC inhibitor

A potent, cell-permeable histone deacetylase (HDAC) inhibitor. Inhibits HDACs in maize, Physarum and chicken. IC50=30 nM for E. tenella HDAC. Displays antiprotozoal and antineoplastic activity. Induces expression of the γ-globin gene in erythroid cells. Shows antimitogenic activities. Induces cell cycle arrest and apoptosis in tumor cells.

Specifications

Specifications

  • Pk:
    1 mg
  • Cat. No.:
    ALEXBML-GR320-0001
  • Protein/peptide name:
    HC Toxin
  • Purity:
    >98%

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.