Specifications
- Pk:0,5 mg
- Seq:02
- Protein/peptide name:Autocamtide-2-related inhibitory Peptide
- Purity:>98%
Specifications
About this item
Cell permeable CaM kinase II inhibitor
Corresponds to Prod. No. ALX-151-029 but this one is N-terminal myristoylated to increase cell permeability.