ContactUsLinkComponent
Mg²⁺/ATP Activating Solution
Mg²⁺/ATP Activating Solution
Catalog # ENZOBMLEW98050100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Mg²⁺/ATP Activating Solution
Catalog # ENZOBMLEW98050100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Pk:
    100 µl
  • Seq:
    02
  • Enzyme name:
    Mg2+/ATP activating solution
Specifications
About this item
This solution contains pre-coupled Mg-ATP for immediate use as an energy source for a range of ATP-dependent reactions (e.g. ubiquitin thioester conjugation, proteasomal degradation, SUMO conjugation).
Liquid. In 50mM TRIS-Cl, pH 7,5, containing 0,1 M ATP and 0,1 M MgCl₂.