Order Entry
Northern Ireland
ContactUsLinkComponent
Human recombinant MKP-1/p38 complex (from insect cells (Sf9))
Human recombinant MKP-1/p38 complex (from insect cells (Sf9))
Catalog # ALEXBML-SE440-0100
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant MKP-1/p38 complex (from insect cells (Sf9))
Catalog # ALEXBML-SE440-0100
Supplier:  Enzo Life Sciences
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Human
  • Protein/peptide name:
    MKP-1/p38 complex

Specifications

About this item

Produced in Sf9 cells.

Approximately 1:1 stoichiometric complex of recombinant human MKP-1 (MAP Kinase Phopshatase-1, DUSP1, CL100) with recombinant human p38α.