About this item
This peptide sequence, KKKVSRSGLYRSPSMPENLNRPR, corresponds to amino acids 206 to 225 of the phosphatase CDC25C, which is a phosphorylation target of Chk1 on Ser216.
- Both Chk1 and Chk2 will phosphorylate this peptide
The addition of three N-terminal lysines allows the peptide to be used in phosphocellulose kinase assays.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Protein/peptide name:Chk 1 and 2 substrate