Order Entry
Northern Ireland
ContactUsLinkComponent
Blocking peptide for Heme oxygenase 1 pAb (Prod. No. BmL-HC3001) Blocking Peptide
Blocking peptide for Heme oxygenase 1 pAb (Prod. No. BmL-HC3001) Blocking Peptide
Catalog # ENZOBMLHP93010100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Blocking peptide for Heme oxygenase 1 pAb (Prod. No. BmL-HC3001) Blocking Peptide
Catalog # ENZOBMLHP93010100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    100 µG
  • Seq:
    02
  • Protein/peptide name:
    Blocking Peptide for Heme oxygenase 1 pAb (Prod. No. BmL-HC3001) blocking peptide
  • Purity:
    >96%

Specifications

About this item

Synthetic peptide corresponding to amino acid residues [12-25] + Cys-NH2 of human heme oxygenase-1.

Complementary peptide to rabbit polyclonal antibody (BML-HC3001) for use in control experiments.

Preincubation of antibody with this peptide abolishes the antibody reactivity with HO-1 protein.