Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pk:1 mg
- Conjugation:Biotin
- Reconstitution instructions:Reconstitute with water to 1mg/ml stock solution. Through the use of either a homogeniser or sonicator, a homogenous solution can be prepared.
- Protein/peptide name:Pam3Cys-Ser-(Lys)4 (Aca-Aca-Biotin) . 2 Trifluoroacetate
- Formulation:Lyophilized
Selective agonist of TLR1/TLR2 complex
Selective, biotin-labelled ligand of Toll-like receptor 1 (TLR1) complexed with TLR2. Cell permeable, water soluble synthetic cationic lipohexapeptide analog of the immunologically active N-terminal portion of bacterial lipoprotein that potently activates monocytes and macrophages. Activates/represses an array of at least 140 genes partly involved in signal transduction and regulation of the immune response.