Order Entry
Northern Ireland
ContactUsLinkComponent
Wash buffer, 3 packs of salts, SignaSure®
Wash buffer, 3 packs of salts, SignaSure®
Catalog # ALEXENZ-33803
Supplier:  Enzo Life Sciences
CAS Number:  
Wash buffer, 3 packs of salts, SignaSure®
Catalog # ALEXENZ-33803
Supplier:  Enzo Life Sciences
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    3 L
  • Description:
    Signasure® wash buffer
  • Seq:
    01
  • Pk Info:
    3× 1 L packs
  • pH value:
    8,0 ±0,2 (solution)

Specifications

About this item

SignaSure® Wash Buffer is formulated for use with alkaline phosphatase-linked detection reagent assay systems.

  • Each packet of salts yields 1 liter of buffer
  • Lyophilized

Dissolve entire contents of packet in 1 liter of distilled or deionized water.