About this item
This kit is a complete assay system to measure protease activity of recombinant caspase-1. Cleavage of a tetrapeptide substrate is monitored colorimetrically at 405 nm or fluorimetrically (Ex. 360 nm/Em. 460 nm).
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Storage conditions:–80 °C (long term)
- Application:Colorimetric detection, fluorescent detection, HTS activity assay
- Size:96 well