Order Entry
Northern Ireland
ContactUsLinkComponent
Stop solution
Stop solution
Catalog # ASSAADI-80-0247
Supplier:  Enzo Life Sciences
CAS Number:  
Stop solution
Catalog # ASSAADI-80-0247
Supplier:  Enzo Life Sciences
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    5 ml
  • Description:
    Stop solution
  • Seq:
    01
  • Storage conditions:
    Ambient

Specifications

About this item

Assay stop solution for AP EIA/ELISA. This is a solution of TRIS-sodium phosphate in water.

  • Liquid in form