- Pk:1 KIT
- Primary antibody reactivity:Human
- Description:SLPI (human), ELISA kit
- Seq:02
- Sensitivity:1 ng/ml (range 10 - 160 ng/ml)
Rapid ELISA kit to quantify SLPI, a secretory leukocyte peptidase inhibitor and unique cancer biomarker.
For the quantitative determination of human SLPI (ALP, Antileukoproteinase) in serum.
The SLPI ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of SLPI in serum with results in just 2.5 hours.