ContactUsLinkComponent
Carboxyfluorescein multi-caspase activity kit
Catalog # ALEXBML-AK117-0001
Supplier:  Enzo Life Sciences
CAS Number:  
Carboxyfluorescein multi-caspase activity kit
Catalog # ALEXBML-AK117-0001
Supplier:  Enzo Life Sciences
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Pk:
    1 Tests
  • Description:
    Carboxyfluorescein multi-caspase activity kit
Specifications
About this item
The Carboxyfluorescein Multi-Caspase Activity Kit detects apoptosis through the covalent binding of a green fluorescent probe to active caspase enzymes in living cells.
  • Fluorescent detection
  • HTS Activity assay
The carboxyfluorescein-labeled caspase inhibitor probe (FAM-VAD-FMK; excitation at 490 nm and emission at 520 nm), comprising a carboxyfluorescein derivative of valylalanylaspartic acid fluoromethyl ketone (VAD-FMK), covalently binds to the reactive cysteine (Cys 285) on the large subunit of active caspase heterodimers. This probe is both cell permeable and non-cytotoxic during the time of the labeling procedure and allows detection of apoptosis by fluorescence microscopy, flow cytometry, and 96-well-plate based fluorometry.
Use of the fluorescence microscope provides a qualitative view of the level of apoptosis induced as compared to the use of a fluorometer or a flow cytometer which will generate quantitative data. The Sulforhodamine Multi-Caspase Activity Kit for Apoptosis Detection is a related kit that contains a sulforhodamine-labeled probe (Ex: 550, Em: 595).
Store the kit at 2-8 °C and protect the FAM-VAD-FMK reagent from light at all times. The 150X FAM-VAD-FMK stock should be stored at -20°C protected from light, this reagent is stable for up to 6 months and may be thawed twice during that time.
Delivery: Kit contains FAM-VAD-FMK Reagent, 10X Wash Buffer, Fixative and Propidium Iodide.