Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:LMP-1
- Antigen symbol:LMP1
- Clonality:Polyclonal
- Clone:D24-G
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Form:Liquid
- Antigen synonyms:EBV|Latent Membrane Protein-1
- Storage buffer:In 20mM TRIS/HCl, pH 8.0, containing 20mg/ml BSA and 0.05% sodium azide.
- Storage temperature:+4°C
- Shipping temperature:Blue Ice Not Frozen
- Immunogen:Synthetic peptide corresponding to the internal region of Epstein-Barr virus, LMP-1 (latent membrane protein-1).
- Purification:Major clone of IgGs obtained from immunoaffinity purified immunoglobulins corresponding to immunogenic peptide.
- Size:500 µl
- Pk:500 µl
Recommended Applications: IHC (FS), IHC (PS)
Species reactivity: Epstein-Barr virus
Type: Primary
Antigen: LMP1
Clonality: Polyclonal
Clone: D24-G
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Epstein-Barr virus