About this item
Type: Primary
Antigen: ESR
Clonality: Monoclonal
Clone: h-151
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- ENZOADISRA1000F
- Antigen symbol:
- ESR
- Antigen name:
- Estrogen Receptor
- Antigen synonyms:
- estrogen receptor alpha delta 4*,5,6,7*/654 isoform|estradiol receptor|nuclear receptor subfamily 3 group A member 1|estrogen receptor|ER-alpha|estrogen receptor alpha delta 3*,4,5,6,7*/819-2 isoform|estrogen receptor alpha delta 4 +49 isoform
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- h-151
- Reactivity:
- Human, Rat
- Host:
- Mouse
- Western blot:
- Yes
- ImmunoChemistry:
- Yes
- ImmunoPrecipitation:
- Yes
- Immunogen:
- Synthetic peptide corresponding to the sequence of human estrogen receptor, conjugated to KLH.
- Form:
- Liquid
- Storage buffer:
- PBS, pH 7,2 containing 50% glycerol and 0,09% sodium azide
- Storage temperature:
- –20 °C
- Shipping temperature:
- Shipped on blue ice