Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-TAPBP Rabbit Polyclonal Antibody
 
Anti-TAPBP Rabbit Polyclonal Antibody

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Catalog No:
  • ENZOADICSA630F
  • ENZOADICSA630D
  • Antigen symbol:
  • TAPBP
  • TAPBP
  • Antigen name:
  • Tapasin
  • Tapasin
  • Antigen synonyms:
  • DADB-159G18.7|TAPA|TPN|TPSN|NGS17
  • DADB-159G18.7|TAPA|TPN|TPSN|NGS17
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic peptide corresponding to aa 2-20 of human tapasin with a cysteine residue added
  • Synthetic peptide corresponding to aa 2-20 of human tapasin with a cysteine residue added
  • Amino acid number:
  • 2 to 20
  • 2 to 20
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Purified by Protein A affinity chromatography
  • Purified by Protein A affinity chromatography
  • Storage buffer:
  • PBS containing 50% glycerol and 0.09% sodium azide
  • PBS containing 50% glycerol and 0.09% sodium azide
  • Storage temperature:
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

 

Frequently Bought Together