Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-ADAM17 Rabbit Polyclonal Antibody
Anti-ADAM17 Rabbit Polyclonal Antibody
Catalog # ENZOADI905249100
Supplier:  ENZO LIFE SCIENCES
Anti-ADAM17 Rabbit Polyclonal Antibody
Catalog # ENZOADI905249100
Supplier:  ENZO LIFE SCIENCES

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ADAM Metallopeptidase Domain 17
  • Antigen symbol:
    ADAM17
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Seq:
    02
  • Epitope:
    near the C-Terminus
  • Form:
    Liquid
  • Antigen synonyms:
    TACE|Snake venom-like protease|ADAM18|CSVP|TNF-alpha-converting enzyme|CD156B|CD156b antigen|TNF-alpha convertase|A disintegrin and metalloproteinase domain 17|NISBD|NISBD1
  • Storage buffer:
    PBS containing 0,02% sodium azide.
  • Storage temperature:
    4 °C
  • Shipping temperature:
    Blue ice not frozen
  • Immunogen:
    Synthetic peptide corresponding to the sequence near the C-terminus of human ADAM17.
  • Purification:
    purified by peptide affinity chromatography
  • Pk:
    100 µG

Specifications

About this item

TNF-alpha converting enzyme (TACE) is a membrane-bound metalloprotease-disintegrin in the family of mammalian ADAM (for a disintegrin and metalloprotease), and is responsible for the cleavage of membrane-bound precursor TNF-alpha to its mature, secreted form. TACE also processes other cell surface proteins, including TNF receptor, TGFalpha, the L-selectin adhesion molecule, and alpha-cleavage of amyloid protein precursor (APP).

Type: Primary
Antigen: ADAM17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-Terminus
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat