Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Complement Factor H
- Antigen symbol:CFH
- Clonality:Monoclonal
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Seq:03
- Form:Liquid, in 0,01 M phosphate buffer, pH 7,4, containing 0,5 M sodium chloride and 15 mM sodium azide
- Purification:Protein A/G-affinity purified
- Pk:1 mg
Factor H is a regulatory factor of the alternative pathway of complement activation. Factor H is a glycoprotein consisting of a single polypeptide chain with a molecular mass of 150kDa.
- For ELISA, Flow Cytometry, Western Blot
Recognizes human complement factor H. Detects a band of ~150kDa by Western blot.
Type: Primary
Antigen: COMPLEMENT FACTOR H
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 ?
Reactivity: Human