Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Proteasome 19S Rpn12/S14 Subunit
- Antigen symbol:Proteasome 19S Rpn12/S14
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:A. thaliana
- Western blot:Yes
- Seq:02
- Form:Liquid, serum
- Pk:25 µl
Specifications
About this item
Host: Rabbit
- For Western Blot
Recognizes Arabidopsis proteasome 26S (19S regulator complex) (Rpn12a/S14 subunit).
Type: Primary
Antigen: PROTEASOME 19S RPN12/S14 SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Plant (Arabidopsis Thaliana)